SY3H3D681722 Synthonix Corporation

6-NITRO-2-PHENYL-1H-BENZO[D]IMIDAZOLE

  • Cas Number : 1571-85-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57356.html

Safety Information

MSDS https://synthonix.com/57356.html
Availability details for
Quantity
  • Availability
  • Add to Cart