SY3H3D681724 Synthonix Corporation

1-(Naphthalen-1-yl)propan-1-amine hydrochloride

  • MDL number No MDL found
  • Cas Number : 149854-36-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57414.html

Safety Information

MSDS https://synthonix.com/57414.html
Availability details for
Quantity
  • Availability
  • Add to Cart