SY3H3D681726 Synthonix Corporation

(S)-1-(Naphthalen-2-yl)propan-1-amine hydrochloride

  • Cas Number : 1810074-89-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57416.html

Safety Information

MSDS https://synthonix.com/57416.html
Availability details for
Quantity
  • Availability
  • Add to Cart