SY3H3D681727 Synthonix Corporation

4-nitrophenyl 2-(pyridin-2-yldisulfanyl)ethyl carbonate

  • MDL number No MDL found
  • Cas Number : 874302-76-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/57561.html

Safety Information

MSDS https://synthonix.com/57561.html
Availability details for
Quantity
  • Availability
  • Add to Cart