SY3H3D681728 Synthonix Corporation

(6R,7R)-4-Nitrobenzyl 7-amino-3-chloro-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate hydrochloride

  • MDL number No MDL found
  • Cas Number : 53483-70-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/57562.html

Safety Information

MSDS https://synthonix.com/57562.html
Availability details for
Quantity
  • Availability
  • Add to Cart