SY3H3D68172D Synthonix Corporation

5-(Naphthalen-1-yl)-1H-pyrrole-2-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 368211-43-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/58141.html

Safety Information

MSDS https://synthonix.com/58141.html
Availability details for
Quantity
  • Availability
  • Add to Cart