SY3H3D68172F Synthonix Corporation

Nonyl(phenyl)sulfane

  • MDL number No MDL found
  • Cas Number : 56358-04-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/58482.html

Safety Information

MSDS https://synthonix.com/58482.html
Availability details for
Quantity
  • Availability
  • Add to Cart