SY3H3D681730 Synthonix Corporation

(3-(Naphthalen-2-yl)phenyl)boronic acid

  • MDL number No MDL found
  • Cas Number : 870774-29-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/58613.html

Safety Information

MSDS https://synthonix.com/58613.html
Availability details for
Quantity
  • Availability
  • Add to Cart