SY3H3D681731 Synthonix Corporation

3-(Naphthalen-2-yloxy)propyl acrylate

  • MDL number MFCD28388316
  • Cas Number : 1642857-58-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form White to light-yellow powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/58681.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/58681.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart