SY3H3D681732 Synthonix Corporation

4-(4-(4-Nitrophenyl)-1-piperazinyl)phenol

  • MDL number No MDL found
  • Cas Number : 112559-81-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58693.html

Safety Information

MSDS https://synthonix.com/58693.html
Availability details for
Quantity
  • Availability
  • Add to Cart