SY3H3D681733 Synthonix Corporation

(R)-2-((4-Nitrophenethyl)amino)-1-phenylethanol

  • MDL number No MDL found
  • Cas Number : 223673-34-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/58703.html

Safety Information

MSDS https://synthonix.com/58703.html
Availability details for
Quantity
  • Availability
  • Add to Cart