SY3H3D681734 Synthonix Corporation

(R)-2-((4-Nitrophenethyl)amino)-1-phenylethanol hydrochloride

  • Cas Number : 521284-21-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/58882.html

Safety Information

MSDS https://synthonix.com/58882.html
Availability details for
Quantity
  • Availability
  • Add to Cart