SY3H3D681739 Synthonix Corporation

Propan-2-amine hydrochloride

  • MDL number MFCD00050705
  • Cas Number : 15572-56-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/39142.html

Safety Information

signalword Warning
pcodes P202, P261, P280
MSDS https://synthonix.com/39142.html
pictograms GHS07
hstatements H315, H319
Availability details for
Quantity
  • Availability
  • Add to Cart