SY3H3D68173C Synthonix Corporation

2-(Propan-2-ylidene)hydrazinecarboxamide

  • MDL number No MDL found
  • Cas Number : 110-20-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/39156.html

Safety Information

MSDS https://synthonix.com/39156.html
Availability details for
Quantity
  • Availability
  • Add to Cart