SY3H3D6817E0 Synthonix Corporation

2-Phenylpropan-1-amine hydrochloride

  • MDL number No MDL found
  • Cas Number : 20388-87-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/39584.html

Safety Information

MSDS https://synthonix.com/39584.html
Availability details for
Quantity
  • Availability
  • Add to Cart