SY3H3D6817E1 Synthonix Corporation

1-(3-Pyridyl)-3-(dimethylamino)-2-propen-1-one

  • MDL number No MDL found
  • Cas Number : 55314-16-4
  • Molecular Weight : 176.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerator
Physical Form Crystal - Powder / Colour: White - Pale yellow - Yellow red - Brown
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/39599.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39599.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart