SY3H3D6817E5 Synthonix Corporation

Propyl 4-methylbenzenesulfonate

  • MDL number No MDL found
  • Cas Number : 599-91-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/39616.html

Safety Information

MSDS https://synthonix.com/39616.html
Availability details for
Quantity
  • Availability
  • Add to Cart