SY3H3D6817E6 Synthonix Corporation

(+)-2-Pinene

  • Cas Number : 7785-70-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/39621.html

Safety Information

MSDS https://synthonix.com/39621.html
Availability details for
Quantity
  • Availability
  • Add to Cart