SY3H3D6817ED Synthonix Corporation

Potassium 2-(4-hydroxyphenyl)-2-((4-methoxy-4-oxobut-2-en-2-yl)amino)acetate

  • MDL number No MDL found
  • Cas Number : 69416-61-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/39717.html

Safety Information

MSDS https://synthonix.com/39717.html
Availability details for
Quantity
  • Availability
  • Add to Cart