SY3H3D6817F8 Synthonix Corporation

1H-Pyrazole-3,5-diamine

  • Cas Number : 16082-33-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/39867.html

Safety Information

MSDS https://synthonix.com/39867.html
Availability details for
Quantity
  • Availability
  • Add to Cart