SY3H3D6817FA Synthonix Corporation

sodium propylsulfamoylazanide

  • MDL number MFCD28964001
  • Cas Number : 1642873-03-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/39890.html

Safety Information

MSDS https://synthonix.com/39890.html
Availability details for
Quantity
  • Availability
  • Add to Cart