SY3H3D681803 Synthonix Corporation

N-Isopropylhydroxylamine hydrochloride

  • MDL number MFCD00012599
  • Cas Number : 50632-53-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: Colorless - White - Almost white
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/39934.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39934.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart