SY3H3D68180E Synthonix Corporation

Propyl carbamate

  • MDL number No MDL found
  • Cas Number : 627-12-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/40264.html

Safety Information

MSDS https://synthonix.com/40264.html
Availability details for
Quantity
  • Availability
  • Add to Cart