SY3H3D681811 Synthonix Corporation

1H-Pyrazole-4-carbonitrile hydrochloride

  • MDL number No MDL found
  • Cas Number : 1416351-94-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/40318.html

Safety Information

MSDS https://synthonix.com/40318.html
Availability details for
Quantity
  • Availability
  • Add to Cart