SY3H3D681814 Synthonix Corporation

Pyrazine-2,6-diamine hydrochloride

  • MDL number No MDL found
  • Cas Number : 1370411-48-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/40325.html

Safety Information

MSDS https://synthonix.com/40325.html
Availability details for
Quantity
  • Availability
  • Add to Cart