SY3H3D681821 Synthonix Corporation

Pent-4-ene-1-sulfonamide

  • MDL number No MDL found
  • Cas Number : 245368-50-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/40414.html

Safety Information

MSDS https://synthonix.com/40414.html
Availability details for
Quantity
  • Availability
  • Add to Cart