SY3H3D681822 Synthonix Corporation

Pentanehydrazide

  • Cas Number : 38291-82-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/40425.html

Safety Information

MSDS https://synthonix.com/40425.html
Availability details for
Quantity
  • Availability
  • Add to Cart