SY3H3D681828 Synthonix Corporation

1H-Pyrrole-3-carbonitrile

  • MDL number No MDL found
  • Cas Number : 7126-38-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/40700.html

Safety Information

MSDS https://synthonix.com/40700.html
Availability details for
Quantity
  • Availability
  • Add to Cart