SY3H3D681829 Synthonix Corporation

Pyrimidine-4-carbaldehyde

  • Cas Number : 2435-50-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/40755.html

Safety Information

MSDS https://synthonix.com/40755.html
Availability details for
Quantity
  • Availability
  • Add to Cart