SY3H3D68182D Synthonix Corporation

Pyrimidine-4-carboxamide

  • MDL number No MDL found
  • Cas Number : 28648-86-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/40959.html

Safety Information

MSDS https://synthonix.com/40959.html
Availability details for
Quantity
  • Availability
  • Add to Cart