SY3H3D681834 Synthonix Corporation

1H-Pyrazole-3,5-dicarboxamide

  • MDL number No MDL found
  • Cas Number : 1397683-79-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/41107.html

Safety Information

MSDS https://synthonix.com/41107.html
Availability details for
Quantity
  • Availability
  • Add to Cart