SY3H3D68183A Synthonix Corporation

Pyridine-2,4,5-triamine

  • MDL number No MDL found
  • Cas Number : 23244-87-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/41207.html

Safety Information

MSDS https://synthonix.com/41207.html
Availability details for
Quantity
  • Availability
  • Add to Cart