SY3H3D68183D Synthonix Corporation

4-Nitroisoindolin-1-one

  • MDL number No MDL found
  • Cas Number : 366452-97-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/47897.html

Safety Information

MSDS https://synthonix.com/47897.html
Availability details for
Quantity
  • Availability
  • Add to Cart