SY3H3D681846 Synthonix Corporation

(S)-Piperidin-3-amine dihydrochloride

  • MDL number No MDL found
  • Cas Number : 334618-07-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/41385.html

Safety Information

MSDS https://synthonix.com/41385.html
Availability details for
Quantity
  • Availability
  • Add to Cart