SY3H3D68184B Synthonix Corporation

Potassium 6-hydroxyhexanoate

  • MDL number No MDL found
  • Cas Number : 21810-34-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/41406.html

Safety Information

MSDS https://synthonix.com/41406.html
Availability details for
Quantity
  • Availability
  • Add to Cart