SY3H3D681852 Synthonix Corporation

Potassium (3-chlorophenyl)trifluoroborate

  • MDL number MFCD04115754
  • Cas Number : 411206-75-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form White to off-white powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41413.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41413.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart