SY3H3D681856 Synthonix Corporation

Potassium 1-cyano-3-ethoxy-2,3-dioxopropan-1-ide

  • MDL number No MDL found
  • Cas Number : 92664-05-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/41417.html

Safety Information

MSDS https://synthonix.com/41417.html
Availability details for
Quantity
  • Availability
  • Add to Cart