SY3H3D68185C Synthonix Corporation

5-Isopropylimidazolidine-2,4-dione

  • MDL number No MDL found
  • Cas Number : 16935-34-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/41435.html

Safety Information

MSDS https://synthonix.com/41435.html
Availability details for
Quantity
  • Availability
  • Add to Cart