SY3H3D68185D Synthonix Corporation

3-(1H-Pyrazol-1-yl)propane-1,2-diol

  • MDL number MFCD11846459
  • Cas Number : 98484-49-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/41436.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/41436.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart