SY3H3D681860 Synthonix Corporation

(1-Propyl-1H-pyrazol-4-yl)boronic acid

  • MDL number No MDL found
  • Cas Number : 847818-57-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/41485.html

Safety Information

MSDS https://synthonix.com/41485.html
Availability details for
Quantity
  • Availability
  • Add to Cart