SY3H3D681867 Synthonix Corporation

Isopropyl cyclopropanesulfonate

  • MDL number No MDL found
  • Cas Number : 146475-51-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/41562.html

Safety Information

MSDS https://synthonix.com/41562.html
Availability details for
Quantity
  • Availability
  • Add to Cart