SY3H3D681870 Synthonix Corporation

Pyrazolo[1,5-a]pyrimidin-5(1H)-one

  • MDL number No MDL found
  • Cas Number : 1027534-43-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/42477.html

Safety Information

MSDS https://synthonix.com/42477.html
Availability details for
Quantity
  • Availability
  • Add to Cart