SY3H3D681871 Synthonix Corporation

1H-Pyrazolo[3,4-b]pyridin-3(2H)-one

  • Cas Number : 2942-43-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/42478.html

Safety Information

MSDS https://synthonix.com/42478.html
Availability details for
Quantity
  • Availability
  • Add to Cart