SY3H3D681872 Synthonix Corporation

1H-Pyrazolo[3,4-c]pyridin-3(2H)-one

  • MDL number No MDL found
  • Cas Number : 53975-70-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/42479.html

Safety Information

MSDS https://synthonix.com/42479.html
Availability details for
Quantity
  • Availability
  • Add to Cart