SY3H3D681874 Synthonix Corporation

1H-Pyrrolo[3,2-d]pyrimidine-2,4(3H,5H)-dione

  • MDL number No MDL found
  • Cas Number : 65996-50-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/42485.html

Safety Information

MSDS https://synthonix.com/42485.html
Availability details for
Quantity
  • Availability
  • Add to Cart