SY3H3D6818C2 Synthonix Corporation

5-(4-Pyridyl)-1H-tetrazole

  • MDL number No MDL found
  • Cas Number : 14389-12-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/42508.html

Safety Information

MSDS https://synthonix.com/42508.html
Availability details for
Quantity
  • Availability
  • Add to Cart