SY3H3D6818CD Synthonix Corporation

1,3-Phenylenediboronic acid

  • MDL number No MDL found
  • Cas Number : 4612-28-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/43318.html

Safety Information

MSDS https://synthonix.com/43318.html
Availability details for
Quantity
  • Availability
  • Add to Cart