SY3H3D6818CE Synthonix Corporation

2-Propylthiazole

  • MDL number No MDL found
  • Cas Number : 17626-75-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/43394.html

Safety Information

MSDS https://synthonix.com/43394.html
Availability details for
Quantity
  • Availability
  • Add to Cart