SY3H3D6818D0 Synthonix Corporation

Pyridin-4-ylmethanol hydrobromide

  • MDL number No MDL found
  • Cas Number : 65737-59-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/43429.html

Safety Information

MSDS https://synthonix.com/43429.html
Availability details for
Quantity
  • Availability
  • Add to Cart