SY3H3D6818D9 Synthonix Corporation

Pyrazolo[1,5-a]pyrimidin-3-amine hydrochloride

  • MDL number No MDL found
  • Cas Number : 232600-78-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/43523.html

Safety Information

MSDS https://synthonix.com/43523.html
Availability details for
Quantity
  • Availability
  • Add to Cart